Assay Details
Target Gene Details
Entrez Gene ID: | 100038356 |
Gene Name: | predicted gene 15612 |
Gene Aliases: |
ENSMUSG00000072920, OTTMUSG00000024319 |
Location: |
Chr.6:88842854-88847274 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm15612 | NR_045880.1 | 2 | 359 | |
| AK133241.1 | 2 | 360 | BAE21574.1 |
Target Gene Details
Entrez Gene ID: | 319655 |
Gene Name: | podocalyxin-like 2 |
Gene Aliases: |
D130074J02Rik, PODLX2 |
Location: |
Chr.6:88842557-88875053 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Podxl2 | NM_176973.4 | 7 | 1492 | NP_795947.3 |
| AK051761.1 | 7 | 1695 | ||
| BC033384.1 | 7 | 1492 | AAH33384.1 |

Genomic Map