Assay Details
Target Gene Details
Entrez Gene ID: | 56223 |
Gene Name: | fascin homolog 3, actin-bundling protein, testicular (Strongylocentrotus purpuratus) |
Gene Aliases: |
- |
Location: |
Chr.6:28427870-28439398 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fscn3 | NM_019569.2 | 4 | 1150 | NP_062515.2 |
| AF176024.2 | 4 | 1262 | AAF19797.1 | |
| BC050759.1 | 4 | 1150 | AAH50759.1 |
More Information
Set Membership: |
|

Genomic Map