Assay Details
Target Gene Details
Entrez Gene ID: | 53379 |
Gene Name: | heterogeneous nuclear ribonucleoprotein A2/B1 |
Gene Aliases: |
9130414A06Rik, Hnrpa2, Hnrpa2b1, hnrnp-A |
Location: |
Chr.6:51460434-51470492 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hnrnpa2b1 | NM_016806.3 | NP_058086.2 | ||
| NM_182650.4 | NP_872591.1 | |||
| NR_104468.1 | ||||
| AF073993.1 | AAC26867.1 | |||
| AF406651.2 | AAK98601.2 | |||
| AK049771.1 | ||||
| AK089012.1 | BAC40700.1 | |||
| AK137214.1 | BAE23274.1 | |||
| BB627924.1 | ||||
| BC059107.1 | AAH59107.1 | |||
| BC141253.1 | ||||
| BC145588.1 |

Genomic Map