Assay Details
Target Gene Details
Entrez Gene ID: | 100043861 |
Gene Name: | killer cell lectin-like receptor subfamily B member 1 |
Gene Aliases: |
4930431A04Rik, Gm4696, Klrb1g, Klrb6, Ly-55, Ly55, NKR-P1G, Nkrp-1e, Nkrp1g |
Location: |
Chr.6:128706093-128764561 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Klrb1 | NM_001099918.1 | 1 | 42 | NP_001093388.1 |
| DQ113419.1 | 1 | 42 | ||
| HQ713436.1 | 1 | 42 | ||
| JF429832.1 | 1 | 55 |
More Information
Set Membership: |
|

Genomic Map