Assay Details
Target Gene Details
Entrez Gene ID: | 98758 |
Gene Name: | heterogeneous nuclear ribonucleoprotein F |
Gene Aliases: |
4833420I20Rik, AA407306, Hnrpf |
Location: |
Chr.6:117900340-117925622 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hnrnpf | NM_001166427.1 | NP_001159899.1 | ||
| NM_001166428.1 | NP_001159900.1 | |||
| NM_001166429.1 | NP_001159901.1 | |||
| NM_001166430.1 | NP_001159902.1 | |||
| NM_001166431.1 | NP_001159903.1 | |||
| NM_001166432.1 | NP_001159904.1 | |||
| NM_133834.2 | NP_598595.1 | |||
| AK076478.1 | BAC36361.1 | |||
| AK177225.1 | ||||
| AK188037.1 | ||||
| AK196656.1 | ||||
| AU079872.1 | ||||
| BC018185.1 | AAH18185.1 | |||
| BC025481.1 | AAH25481.1 | |||
| BC089313.1 | AAH89313.1 | |||
| BY774552.1 | ||||
| CA539382.1 | ||||
| CF163622.1 | ||||
| CJ048596.1 | ||||
| CJ054614.1 | ||||
| CJ094336.1 | ||||
| CK389060.1 |
More Information
Set Membership: |
|

Genomic Map