Assay Details
Target Gene Details
Entrez Gene ID: | 330267 |
Gene Name: | thrombospondin, type I, domain containing 7A |
Gene Aliases: |
Gm837 |
Location: |
Chr.6:12311608-12749253 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Thsd7a | NM_001164805.1 | 5 | 1794 | NP_001158277.1 |
| AK136087.1 | 4 | 725 | BAE22813.1 | |
| AK173072.1 | 5 | 1565 | BAD32350.1 |
More Information
Set Membership: |
|

Genomic Map