Assay Details
Target Gene Details
Entrez Gene ID: | 233545 |
Gene Name: | RIKEN cDNA 2210018M11 gene |
Gene Aliases: |
2310016L09Rik, AU019521, AU019557, Emsy |
Location: |
Chr.7:98587137-98658289 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 2210018M11Rik | NM_172280.2 | NP_758484.2 | ||
| AK008751.1 | ||||
| AK140537.1 | BAE24417.1 | |||
| AK158499.1 | 1 | 1213 | ||
| BC020109.1 | AAH20109.1 | |||
| BC089304.1 | AAH89304.1 |
More Information
Set Membership: |
|

Genomic Map