Assay Details
Target Gene Details
Entrez Gene ID: | 233545 |
Gene Name: | RIKEN cDNA 2210018M11 gene |
Gene Aliases: |
2310016L09Rik, AU019521, AU019557, Emsy |
Location: |
Chr.7:98587137-98658289 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 2210018M11Rik | NM_172280.2 | NP_758484.2 | ||
| AK032985.1 | BAC28113.2 | |||
| AK137691.1 | 3 | 469 | ||
| BC089304.1 | AAH89304.1 |
More Information
Set Membership: |
|

Genomic Map