Assay Details
Target Gene Details
Entrez Gene ID: | 71660 |
Gene Name: | retinoic acid receptor responder (tazarotene induced) 2 |
Gene Aliases: |
0610007L05Rik, AI303516 |
Location: |
Chr.6:48569696-48572844 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rarres2 | NM_027852.2 | NP_082128.1 | ||
| AK002298.1 | BAB21997.1 | |||
| BC038914.1 | AAH38914.1 |

Genomic Map