Assay Details
Target Gene Details
Entrez Gene ID: | 243371 |
Gene Name: | leucine rich repeat containing 61 |
Gene Aliases: |
BC027309 |
Location: |
Chr.6:48554779-48570722 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lrrc61 | NM_001110160.1 | 2 | 2171 | NP_001103630.1 |
| NM_177736.3 | 3 | 2400 | NP_808404.2 | |
| AK143222.1 | 3 | 2403 | BAE25315.1 | |
| AK170792.1 | 2 | 2161 | ||
| AK171463.1 | 2 | 2166 | BAE42469.1 | |
| BC027309.1 | 4 | 2203 | AAH27309.1 | |
| BP765942.1 | 1 | 533 |
Target Gene Details
Entrez Gene ID: | 71660 |
Gene Name: | retinoic acid receptor responder (tazarotene induced) 2 |
Gene Aliases: |
0610007L05Rik, AI303516 |
Location: |
Chr.6:48569696-48572844 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rarres2 | NM_027852.2 | NP_082128.1 | ||
| AK002298.1 | BAB21997.1 | |||
| BC038914.1 | AAH38914.1 |

Genomic Map