Assay Details
Target Gene Details
Entrez Gene ID: | 56372 |
Gene Name: | RIKEN cDNA 1110004F10 gene |
Gene Aliases: |
Smacp, Smap, sid2057, sid2057p |
Location: |
Chr.7:116093380-116105210 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1110004F10Rik | NM_019772.2 | NP_062746.1 | ||
| AB025407.1 | BAA84692.1 | |||
| AK003415.1 | BAB22777.1 | |||
| AK144059.1 | BAE25676.1 | |||
| AK167571.1 | BAE39634.1 | |||
| AK168206.1 | BAE40166.1 | |||
| AK169309.1 | BAE41064.1 | |||
| AK170172.1 | ||||
| BC072575.1 | AAH72575.1 |

Genomic Map