Assay Details
Target Gene Details
Entrez Gene ID: | 66087 |
Gene Name: | ER membrane protein complex subunit 3 |
Gene Aliases: |
0610039A15Rik, AI225901, AW260416, Pob, Tmem111 |
Location: |
Chr.6:113514887-113531638 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Emc3 | NM_175101.3 | 6 | 689 | NP_780310.1 |
| AK020058.1 | 6 | 660 | ||
| AK075595.1 | 6 | 690 | BAC35844.1 | |
| AK167819.1 | 6 | 538 | BAE39843.1 | |
| AK200024.1 | 3 | 142 | ||
| AK210449.1 | ||||
| AK218757.1 | ||||
| BC004641.1 | 6 | 685 | AAH04641.1 |

Genomic Map