Assay Details
Target Gene Details
Entrez Gene ID: | 68487 |
Gene Name: | transmembrane protein 140 |
Gene Aliases: |
1110007F12Rik |
Location: |
Chr.6:34863106-34874946 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem140 | NM_197986.2 | 1 | 133 | NP_932103.1 |
| AK030532.1 | 1 | 134 | BAC27007.1 | |
| AK085154.1 | 1 | 56 | ||
| AK165621.1 | 1 | 136 | ||
| BC023802.1 | 1 | 147 | AAH23802.1 |

Genomic Map