Assay Details
Target Gene Details
Entrez Gene ID: | 69938 |
Gene Name: | secernin 1 |
Gene Aliases: |
2810019K23Rik, 6330535A03Rik, AI852905, SES1, mKIAA0193 |
Location: |
Chr.6:54472300-54566499 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Scrn1 |
Target Gene Details
Entrez Gene ID: | 330319 |
Gene Name: | WAS/WASL interacting protein family, member 3 |
Gene Aliases: |
9030621G03, BQ952480 |
Location: |
Chr.6:54429218-54503768 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Wipf3 | NM_001167860.1 | NP_001161332.1 | ||
| NM_001167861.1 | NP_001161333.1 |
More Information
Set Membership: |
|

Genomic Map