Assay Details
Target Gene Details
Entrez Gene ID: | 73833 |
Gene Name: | family with sequence similarity 98, member C |
Gene Aliases: |
1110006G06Rik, AI836081, AV026483, B230110F21Rik |
Location: |
Chr.7:29152510-29156359 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam98c | NM_001146023.1 | NP_001139495.1 | ||
| AK003496.1 | BAB22821.1 | |||
| AK080790.1 | ||||
| AK138395.1 | ||||
| AK166139.1 | ||||
| AK171108.1 | ||||
| BC023421.1 | AAH23421.1 |
More Information
Set Membership: |
|

Genomic Map