Assay Details
Target Gene Details
Entrez Gene ID: | 73833 |
Gene Name: | family with sequence similarity 98, member C |
Gene Aliases: |
1110006G06Rik, AI836081, AV026483, B230110F21Rik |
Location: |
Chr.7:29152510-29156359 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam98c | NM_001146023.1 | 7 | 845 | NP_001139495.1 |
| AK003496.1 | 6 | 769 | BAB22821.1 | |
| AK080790.1 | 6 | 937 | ||
| AK138395.1 | 6 | 1748 | ||
| AK166139.1 | 6 | 1744 | ||
| AK171108.1 | 5 | 1211 | ||
| BC023421.1 | 4 | 463 | AAH23421.1 |
Target Gene Details
Entrez Gene ID: | 233046 |
Gene Name: | RAS guanyl releasing protein 4 |
Gene Aliases: |
- |
Location: |
Chr.7:29134933-29153961 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rasgrp4 | NM_001174155.1 | 17 | 4083 | NP_001167626.1 |
| NM_145149.4 | 17 | 4068 | NP_660131.2 | |
| NR_045676.1 | 17 | 4046 | ||
| AK089282.1 | 17 | 4015 | BAC40827.1 |
More Information
Set Membership: |
|

Genomic Map