Assay Details
Target Gene Details
Entrez Gene ID: | 67928 |
Gene Name: | ATP-binding cassette, sub-family A (ABC1), member 14 |
Gene Aliases: |
1700110B15Rik, 4930539G24Rik |
Location: |
Chr.7:120203895-120325352 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abca14 | NM_026458.4 | 3 | 376 | NP_080734.3 |
| AK007156.1 | 3 | 337 | BAB24875.1 | |
| AK015996.1 | 3 | 333 | BAB30074.1 | |
| AK016294.1 | 3 | 331 | ||
| AK029665.1 | 3 | 380 | BAC26555.1 | |
| AY243470.1 | 3 | 376 | AAP73044.1 | |
| BC116805.1 | 3 | 307 | ||
| BC116809.1 | 3 | 307 |
More Information
Set Membership: |
|

Genomic Map