Assay Details
Target Gene Details
Entrez Gene ID: | 101185 |
Gene Name: | protection of telomeres 1A |
Gene Aliases: |
1500031H18Rik, AI851169, Pot1 |
Location: |
Chr.6:25743735-25809277 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pot1a | NM_133931.4 | 12 | 1493 | NP_598692.1 |
| AK036052.1 | 12 | 1494 | BAC29288.1 | |
| AK040864.1 | 12 | 1486 | BAC30723.1 | |
| AK085041.1 | 12 | 1484 | BAC39349.1 | |
| AK137376.1 | 12 | 1463 | BAE23333.1 | |
| BC016121.1 | 12 | 1455 | AAH16121.1 |
More Information
Set Membership: |
|

Genomic Map