Assay Details
Target Gene Details
Entrez Gene ID: | 101185 |
Gene Name: | protection of telomeres 1A |
Gene Aliases: |
1500031H18Rik, AI851169, Pot1 |
Location: |
Chr.6:25743735-25809277 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pot1a | NM_133931.4 | NP_598692.1 | ||
| AK036052.1 | BAC29288.1 | |||
| AK040864.1 | BAC30723.1 | |||
| BC016121.1 | AAH16121.1 |
More Information
Set Membership: |
|

Genomic Map