Assay Details
Target Gene Details
Entrez Gene ID: | 69938 |
Gene Name: | secernin 1 |
Gene Aliases: |
2810019K23Rik, 6330535A03Rik, AI852905, SES1, mKIAA0193 |
Location: |
Chr.6:54472300-54566499 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Scrn1 | NM_027268.2 | NP_081544.1 | ||
| AK012765.1 | BAB28453.1 | |||
| AK032573.1 | BAC27931.1 | |||
| AK050481.1 | BAC34280.1 | |||
| AK082953.1 | BAC38706.1 | |||
| AK089968.1 | ||||
| AK129084.1 | BAC97894.1 | |||
| AK145904.1 | BAE26739.1 | |||
| BC023889.1 | AAH23889.1 |

Genomic Map