Assay Details
Target Gene Details
Entrez Gene ID: | 55961 |
Gene Name: | solute carrier family 13 (sodium/sulfate symporters), member 1 |
Gene Aliases: |
AI314066, AI987826, NaSi-1, Nas1 |
Location: |
Chr.6:24088283-24168119 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc13a1 | NM_019481.2 | NP_062354.2 | ||
| AF199365.1 | AAF64648.1 | |||
| AF199366.1 | AAF64649.1 | |||
| AK019876.1 | 3 | 369 | ||
| AK128899.1 | ||||
| BC049981.1 | AAH49981.1 |
More Information
Set Membership: |
|

Genomic Map