Assay Details
Target Gene Details
Entrez Gene ID: | 434234 |
Gene Name: | RIKEN cDNA 2610020H08 gene |
Gene Aliases: |
- |
Location: |
Chr.7:119794130-119882695 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 2610020H08Rik | NM_001004187.1 | NP_001004187.1 | ||
| NM_028129.2 | NP_082405.1 | |||
| AK011486.1 | ||||
| AK084640.1 | ||||
| BC053445.1 | AAH53445.1 |
More Information
Set Membership: |
|

Genomic Map