Assay Details
Target Gene Details
Entrez Gene ID: | 243755 |
Gene Name: | solute carrier family 13 (sodium/sulfate symporters), member 4 |
Gene Aliases: |
9630060C05Rik, SUT-1, SUT1 |
Location: |
Chr.6:35267953-35308126 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc13a4 | NM_172892.3 | 2 | 836 | NP_766480.1 |
| AK036354.1 | 2 | 838 | BAC29395.1 | |
| AK038937.1 | 2 | 840 | ||
| AK139428.1 | 2 | 839 | BAE24009.1 | |
| BC089161.1 | 2 | 825 | AAH89161.1 |
More Information
Set Membership: |
|

Genomic Map