Assay Details
Target Gene Details
Entrez Gene ID: | 52440 |
Gene Name: | Tax1 (human T cell leukemia virus type I) binding protein 1 |
Gene Aliases: |
1200003J11Rik, 1700069J21Rik, AA930106, D6Ertd404e, D6Ertd772e, T6bp, TXBP151 |
Location: |
Chr.6:52713575-52766780 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tax1bp1 | NM_025816.3 | NP_080092.2 | ||
| AK004574.1 | BAB23383.1 | |||
| AK146076.1 | BAE26880.1 | |||
| BC014798.1 | AAH14798.1 |

Genomic Map