Assay Details
Target Gene Details
Entrez Gene ID: | 116732 |
Gene Name: | testis specific gene A13 |
Gene Aliases: |
1700023D02Rik, Vme1 |
Location: |
Chr.6:30896980-30925154 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tsga13 | NM_054073.2 | NP_473414.1 | ||
| AF294348.1 | AAK97090.1 | |||
| AK006265.1 | BAB24491.1 | |||
| BC125597.1 | ||||
| BC125599.1 |

Genomic Map