Assay Details
Target Gene Details
Entrez Gene ID: | 19694 |
Gene Name: | regenerating islet-derived 3 alpha |
Gene Aliases: |
AV063448 |
Location: |
Chr.6:78380709-78383839 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Reg3a | NM_011259.1 | NP_035389.1 | ||
| AK008050.1 | BAB25430.1 | |||
| AK008140.1 | BAB25488.1 | |||
| AK008470.1 | BAB25687.1 | |||
| BC119153.1 | ||||
| D63356.1 | BAA18925.1 | |||
| D63357.1 | BAA18926.1 |

Genomic Map