Assay Details
Target Gene Details
Entrez Gene ID: | 18489 |
Gene Name: | regenerating islet-derived 3 beta |
Gene Aliases: |
HIP, PAP1, Pap, REG-III |
Location: |
Chr.6:78370885-78373466 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Reg3b | NM_011036.1 | NP_035166.1 | ||
| BC027525.1 | AAH27525.1 | |||
| D63359.1 | BAA18928.1 |

Genomic Map