Assay Details
Target Gene Details
Entrez Gene ID: | 381760 |
Gene Name: | single-stranded DNA binding protein 1 |
Gene Aliases: |
2810480P10Rik, G630031O20Rik, MtSSB, mtDBP |
Location: |
Chr.6:40471415-40481825 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ssbp1 | NM_001286663.1 | NP_001273592.1 | ||
| NM_001286664.1 | NP_001273593.1 | |||
| NM_028358.2 | NP_082634.1 | |||
| NM_212468.3 | NP_997633.1 | |||
| AK013425.1 | BAB28850.1 | |||
| AK160556.1 | ||||
| AK187390.1 | ||||
| BC028648.1 | AAH28648.1 | |||
| BC061183.1 | AAH61183.1 |
More Information
Set Membership: |
|

Genomic Map