Assay Details
Target Gene Details
Entrez Gene ID: | 21391 |
Gene Name: | thromboxane A synthase 1, platelet |
Gene Aliases: |
CYP5, CYP5A1, THAS, TS, TXAS, TXS |
Location: |
Chr.6:38918467-39084590 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 10 - Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbxas1 | NM_011539.3 | NP_035669.3 | ||
| AK085414.1 | 7 | 1179 | ||
| AK139479.1 | BAE24030.1 | |||
| BC029014.1 | AAH29014.1 | |||
| L18868.1 | AAB39850.1 |
More Information
Set Membership: |
|

Genomic Map