Assay Details
Target Gene Details
Entrez Gene ID: | 19695 |
Gene Name: | regenerating islet-derived 3 gamma |
Gene Aliases: |
AI449515 |
Location: |
Chr.6:78466268-78468874 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Reg3g | NM_011260.1 | NP_035390.1 | ||
| AK008446.1 | BAB25674.1 | |||
| AK008608.1 | BAB25774.1 | |||
| BC061139.1 | AAH61139.1 | |||
| D63361.1 | BAA18930.1 |
More Information
Set Membership: |
|

Genomic Map