Assay Details
Target Gene Details
Entrez Gene ID: | 53620 |
Gene Name: | vesicle-associated membrane protein 5 |
Gene Aliases: |
AF119384, Camp |
Location: |
Chr.6:72368049-72380468 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vamp5 | NM_001080742.2 | NP_001074211.1 | ||
| NM_016872.4 | NP_058568.1 | |||
| AF035643.1 | AAC95282.1 | |||
| AK009266.1 | BAB26182.1 | |||
| AK145137.1 | BAE26254.1 | |||
| BC086925.1 | AAH86925.1 | |||
| BY709062.1 |

Genomic Map