Assay Details
Target Gene Details
Entrez Gene ID: | 100503036 |
Gene Name: | RIKEN cDNA 1700015G11 gene |
Gene Aliases: |
Ccdc179 |
Location: |
Chr.7:52011680-52015716 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1700015G11Rik | NM_001195601.1 | NP_001182530.1 | ||
| AK005994.1 | ||||
| AV042221.2 | ||||
| AV042559.2 |

Genomic Map