Assay Details
Target Gene Details
Entrez Gene ID: | 64213 |
Gene Name: | suppression of tumorigenicity 7 |
Gene Aliases: |
9430001H04Rik, Fam4a2, HELG, RAY1, SEN4, TSG7 |
Location: |
Chr.6:17692940-17943023 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 13 - Intron 13 |

Genomic Map