Assay Details
Target Gene Details
Entrez Gene ID: | 64213 |
Gene Name: | suppression of tumorigenicity 7 |
Gene Aliases: |
9430001H04Rik, Fam4a2, HELG, RAY1, SEN4, TSG7 |
Location: |
Chr.6:17692940-17943023 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| St7 | NM_001083315.2 | 3 | 601 | NP_001076784.1 |
| NM_001289624.1 | 3 | 525 | NP_001276553.1 | |
| NM_001289625.1 | 3 | 525 | NP_001276554.1 | |
| NM_001289626.1 | 3 | 525 | NP_001276555.1 | |
| NM_001289627.1 | 3 | 259 | NP_001276556.1 | |
| NM_001289629.1 | 3 | 259 | NP_001276558.1 | |
| NM_022332.3 | 3 | 601 | NP_071727.2 | |
| AF234884.1 | 3 | 552 | AAF85947.1 | |
| AF234885.1 | 3 | 552 | AAF85948.1 | |
| AK036248.1 | 3 | 480 | ||
| AK079107.1 | 3 | 602 | ||
| AK132103.1 | 3 | 555 | BAE20983.1 | |
| AK151550.1 | 3 | 556 | BAE30495.1 | |
| AY007792.1 | 3 | 487 | AAG24458.1 | |
| AY007793.1 | 3 | 487 | AAG24459.1 | |
| AY007794.1 | 4 | 516 | AAG24460.1 | |
| AY007795.1 | 4 | 516 | AAG24461.1 | |
| AY007796.1 | 4 | 326 | AAG24462.1 | |
| AY007797.1 | 4 | 326 | AAG24463.1 | |
| AY007798.1 | 3 | 259 | AAG24464.1 | |
| AY007799.1 | 3 | 259 | AAG24465.1 | |
| AY551821.1 | 3 | 252 | AAS59839.1 | |
| AY551822.1 | 3 | 252 | AAS59840.1 | |
| BC024652.1 | 3 | 525 | AAH24652.1 | |
| BC108418.1 | 3 | 545 | AAI08419.1 | |
| CT010245.1 | 3 | 178 | CAJ18453.1 |

Genomic Map