Assay Details
Target Gene Details
Entrez Gene ID: | 319776 |
Gene Name: | transmembrane protein 72 |
Gene Aliases: |
C230095G01Rik |
Location: |
Chr.6:116679103-116716978 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem72 | NM_178768.4 | 4 | 460 | NP_848883.2 |
| AK033856.1 | 4 | 596 | BAC28497.1 | |
| AK049048.1 | 4 | 615 | BAC33524.1 | |
| AK049938.1 | 4 | 573 | BAC33994.1 | |
| AK085603.1 | 4 | 461 | BAC39484.1 | |
| AK141578.1 | 4 | 617 | BAE24746.1 | |
| BC093486.1 | 4 | 603 | ||
| BC116440.1 | 4 | 444 | ||
| BC116441.1 | 4 | 444 |

Genomic Map