Assay Details
Target Gene Details
Entrez Gene ID: | 19240 |
Gene Name: | thymosin, beta 10 |
Gene Aliases: |
Ptmb10, Tb10 |
Location: |
Chr.6:72957347-72958748 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmsb10 | NM_001190327.1 | NP_001177256.1 | ||
| NM_025284.4 | 1 | 38 | NP_079560.1 | |
| AK003178.1 | BAB22622.1 | |||
| AK005512.1 | BAB24091.1 | |||
| AK008557.1 | BAB25742.1 | |||
| AK012361.1 | BAB28189.1 | |||
| AK019212.1 | BAB31603.1 | |||
| AK019232.1 | BAB31614.1 | |||
| AK131711.1 | BAE20774.1 | |||
| AK161987.1 | BAE36668.1 |

Genomic Map