Assay Details
Target Gene Details
Entrez Gene ID: | 232784 |
Gene Name: | Zinc finger protein 212 |
Gene Aliases: |
Znf212, mKIAA3011 |
Location: |
Chr.6:47920568-47932639 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp212 | NM_001145881.1 | NP_001139353.1 | ||
| NM_145576.2 | NP_663551.2 | |||
| CJ054342.1 | 1 | 107 |
More Information
Set Membership: |
|

Genomic Map