Assay Details
Target Gene Details
Entrez Gene ID: | 232784 |
Gene Name: | Zinc finger protein 212 |
Gene Aliases: |
Znf212, mKIAA3011 |
Location: |
Chr.6:47920568-47932639 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp212 | NM_001145881.1 | NP_001139353.1 | ||
| NM_145576.2 | NP_663551.2 | |||
| AK129476.1 | BAC98286.1 | |||
| AK189832.1 | ||||
| BC006693.1 | AAH06693.1 | |||
| BC064742.1 | AAH64742.1 | |||
| BC088737.1 | ||||
| CF915685.1 |
More Information
Set Membership: |
|

Genomic Map