Assay Details
Target Gene Details
Entrez Gene ID: | 76713 |
Gene Name: | RIKEN cDNA 1700039E15 gene |
Gene Aliases: |
Gm19188 |
Location: |
Chr.7:45277331-45288996 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1700039E15Rik | NM_001033176.1 | 5 | 726 | NP_001028348.1 |
| AK133083.1 | 5 | 726 | BAE21503.1 | |
| BC147591.1 | 5 | 823 | ||
| BC147593.1 | 5 | 823 |
More Information
Set Membership: |
|

Genomic Map