Assay Details
Target Gene Details
Entrez Gene ID: | 67442 |
Gene Name: | retinol saturase (all trans retinol 13,14 reductase) |
Gene Aliases: |
0610039N19Rik, C80029, MMT-7, Ppsig |
Location: |
Chr.6:72598628-72607488 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Retsat | NM_026159.4 | 2 | 291 | NP_080435.3 |
| AY704159.1 | 2 | 291 | AAU25836.1 | |
| BC117750.1 | 2 | 286 | ||
| BC117751.1 | 2 | 291 | ||
| EF620365.1 | 2 | 317 |

Genomic Map