Assay Details
Target Gene Details
Entrez Gene ID: | 22147 |
Gene Name: | tubulin, alpha 3B |
Gene Aliases: |
M[a]7, Tuba7 |
Location: |
Chr.6:145615963-145621477 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tuba3b | NM_009449.3 | NP_033475.1 | ||
| CK137160.1 |
More Information
Set Membership: |
|

Genomic Map