Assay Details
Target Gene Details
Entrez Gene ID: | 53857 |
Gene Name: | tubulin, alpha 8 |
Gene Aliases: |
- |
Location: |
Chr.6:121210770-121226097 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tuba8 | NM_017379.1 | NP_059075.1 | ||
| AJ245923.1 | CAB88033.1 | |||
| BC017631.1 | AAH17631.1 |
More Information
Set Membership: |
|

Genomic Map