Assay Details
Target Gene Details
Entrez Gene ID: | 269800 |
Gene Name: | zinc finger protein 384 |
Gene Aliases: |
BB163993, C130073D16Rik, Ciz, NP, Nmp4 |
Location: |
Chr.6:125009110-125037870 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp384 | NM_001252083.1 | NP_001239012.1 | ||
| NM_175557.5 | NP_780766.3 | |||
| AK086185.1 | BAC39626.1 | |||
| AK156026.1 | BAE33551.1 | |||
| AK172476.1 | BAE43028.1 | |||
| BB662626.1 | ||||
| BC064013.1 | AAH64013.1 | |||
| BY261721.1 |
More Information
Set Membership: |
|

Genomic Map