Assay Details
Target Gene Details
Entrez Gene ID: | 19331 |
Gene Name: | RAB19, member RAS oncogene family |
Gene Aliases: |
- |
Location: |
Chr.6:39381428-39390666 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab19 | NM_011226.1 | 3 | 388 | NP_035356.1 |
| AB232613.1 | 2 | 294 | BAF02875.1 | |
| AK017590.1 | 3 | 422 | ||
| BC032936.1 | 3 | 388 | AAH32936.1 | |
| X80473.1 | 2 | 294 | CAA56644.1 |

Genomic Map