Assay Details
Target Gene Details
Entrez Gene ID: | 19349 |
Gene Name: | RAB7, member RAS oncogene family |
Gene Aliases: |
Rab7a |
Location: |
Chr.6:87999106-88045270 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab7 | NM_001293652.1 | NP_001280581.1 | ||
| NM_001293653.1 | NP_001280582.1 | |||
| NM_001293654.1 | NP_001280583.1 | |||
| NM_001293655.1 | NP_001280584.1 | |||
| NM_009005.3 | NP_033031.2 | |||
| AB232599.1 | BAF02861.1 | |||
| AK005004.1 | BAB23738.1 | |||
| AK057671.1 | ||||
| AK135659.1 | ||||
| AK139914.1 | BAE24180.1 | |||
| AK148117.1 | BAE28355.1 | |||
| AK151078.1 | BAE30093.1 | |||
| AK152666.1 | BAE31401.1 | |||
| AK154234.1 | BAE32452.1 | |||
| AK158971.1 | BAE34750.1 | |||
| AK159507.1 | ||||
| AK159928.1 | BAE35490.1 | |||
| AK178672.1 | ||||
| AK183086.1 | ||||
| BC004597.1 | AAH04597.1 | |||
| BC086793.1 | AAH86793.1 | |||
| CT010352.1 | CAJ18560.1 | |||
| X89650.1 | CAA61797.1 |

Genomic Map