Assay Details
Target Gene Details
Entrez Gene ID: | 67442 |
Gene Name: | retinol saturase (all trans retinol 13,14 reductase) |
Gene Aliases: |
0610039N19Rik, C80029, MMT-7, Ppsig |
Location: |
Chr.6:72598628-72607488 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Retsat | NM_026159.4 | 8 | 1312 | NP_080435.3 |
| AK002851.1 | 5 | 664 | BAB22406.2 | |
| AK009103.1 | 4 | 367 | ||
| AY704159.1 | 8 | 1312 | AAU25836.1 | |
| BC011203.1 | 4 | 494 | ||
| BC117750.1 | 8 | 1307 | ||
| BC117751.1 | 8 | 1312 | ||
| EF620365.1 | 8 | 1338 |

Genomic Map