Assay Details
Target Gene Details
Entrez Gene ID: | 50501 |
Gene Name: | prokineticin 2 |
Gene Aliases: |
Bv8, PK2, Prok1 |
Location: |
Chr.6:99711299-99726392 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prok2 | NM_001037539.2 | 1 | 49 | NP_001032628.1 |
| NM_001170419.1 | 1 | 49 | NP_001163890.1 | |
| NM_015768.2 | 1 | 49 | NP_056583.1 | |
| AF182064.1 | 1 | 49 | AAF15259.1 | |
| AF182065.1 | 1 | 49 | AAF15260.1 | |
| AF182066.1 | 1 | 49 | AAF15261.1 | |
| AF487280.1 | 1 | 40 | AAM49572.1 | |
| AK015462.1 | BAB29857.1 | |||
| BB614931.1 | 1 | 41 | ||
| BC116911.1 | ||||
| BC144863.1 | ||||
| BY232101.1 | 1 | 65 |
More Information
Set Membership: |
|

Genomic Map