Assay Details
Target Gene Details
Entrez Gene ID: | 232078 |
Gene Name: | threonine synthase-like 2 (bacterial) |
Gene Aliases: |
BC051244, TSH2 |
Location: |
Chr.6:71128166-71144380 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Thnsl2 | NM_001033929.2 | 5 | 714 | NP_001029101.1 |
| NM_178413.5 | 6 | 782 | NP_848500.3 | |
| AK165463.1 | 5 | 680 | BAE38200.1 | |
| BC025604.1 | 6 | 745 | AAH25604.1 | |
| BC051244.1 | 5 | 676 | AAH51244.1 | |
| BQ042728.1 | 2 | 63 |

Genomic Map