Assay Details
Target Gene Details
Entrez Gene ID: | 320938 |
Gene Name: | transportin 3 |
Gene Aliases: |
5730544L10Rik, C430013M08Rik, C81142, D6Ertd313e, Trn-SR, mKIAA4133 |
Location: |
Chr.6:29540827-29610110 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnpo3 | NM_177296.4 | NP_796270.2 | ||
| AK220184.1 | BAD90369.1 | |||
| BC052756.1 | AAH52756.1 | |||
| BC064646.1 | AAH64646.1 | |||
| BC075678.1 | AAH75678.1 |
More Information
Set Membership: |
|

Genomic Map