Assay Details
Target Gene Details
Entrez Gene ID: | 78118 |
Gene Name: | RIKEN cDNA 4930451I11 gene |
Gene Aliases: |
- |
Location: |
Chr.7:126830468-126831540 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4930451I11Rik | NM_183131.2 | NP_898954.2 | ||
| AK019625.1 | BAC25600.1 | |||
| BC048622.1 | AAH48622.1 |

Genomic Map